[Lys(Me1)4]-Histone H3 (1-21)-GGK,Biotin
Supplier: Anaspec Inc
AS-64355-025
AS-64355-1
103006-528EA
225.08
USD
103006-528
103006-530
[Lys(Me1)4]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is Histone 3 amino acid residues 1 to 21 mono-methylated at Lys-4 with a C-terminal GG linker followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ART-K(Me1)-QTARKSTGGKAPRKQLA-GGK(Biotin)
MW:2737.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...