Buforin
Supplier: Anaspec Inc
AS-61255
103006-482EA
330.31
USD
103006-482
Buforin
Proteins and Peptides
This antimicrobial peptide interacts with phospholipid bilayers and can be efficiently translocated accross the layer with a weak membrane permeabilization activity. The proline hinge in Buforin 2 is responsible for these cell penetrating properties. This class of antibacterial peptides therefore targets intracellular molecules, most probably nucleic acids, without significantly permeting cell membranes.
Sequence:TRSSRAGLQFPVGRVHRLLRK
MW:2434.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...