Human Neuropeptide S
Supplier: Anaspec Inc
AS-61243
103006-450EA
239.69
USD
103006-450
Human Neuropeptide S
Proteins and Peptides
A neuropeptide that plays a role in arousal and fear responses.
Sequence:SFRNGVGTGMKKTSFQRAKS
MW:2187.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...