HIV TAT Cys(Npys) FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
This peptide corresponds to the protein transduction domain of the TAT protein and is synthesized with an activated cysteine residue C(Npys), wherein Npys is 3-Nitro-2-pyridinesulfenyl group and is used for activating S of cysteine and for rapid reaction when a thiol group is introduced. This is synthesized with C(Npys) at the N-terminus for applications requiring specific conjugation reactions and has a fluorophore labeled with FAM having Abs/Em=494/518 nm. This kind of modification has been used to render this peptide as a cell penetrating and carrier peptide applicable in conjugation studies.
Sequence: C(Npys)YGRKKRRQRRR-K(FAM)-NH2
MW: 2302.7 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...