HIV TAT (47-57) Biotin Peptide, Biotin
Supplier: Anaspec Inc
AS-61209
103006-416EA
225.08
USD
103006-416
HIV TAT (47-57) Biotin Peptide, Biotin
Proteins and Peptides
This is a biotinylated HIV-derived cell penetrating TAT peptide . This positively charged biotin-peptide has been used in studies to form a colloidal coat over oligonucleotides-saturated nanoparticles such that TAT-coated nanoparticles when loaded with dense SiRNA molecules could efficiently penetrate a wide variety of human embryonic stem cells.
Sequence: Biotin-YGRKKRRQRRR
MW: 1786.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...