(Arg)9, FAM (Carboxyfluorescein), AnaSpec
Supplier: Anaspec Inc
AS-61207
103006-412EA
454.54
USD
103006-412
(Arg)9, FAM (Carboxyfluorescein), AnaSpec
Proteins and Peptides
(Arg)9 is a cell-permeable peptide used for drug delivery.It can traverse the plasma membrane of eukaryotic cells. This is a fluorescent (FAM)-labeled cell permeable peptide, Abs/Em = 494/521 nm.
Sequence:FAM-RRRRRRRRR
MW:1782 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...