Substance P, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-61201
103006-404EA
423.85
USD
103006-404
Substance P, FAM (Carboxyfluorescein)
Proteins and Peptides
Substance P (SP) is an important neuropeptide belonging to the tachykinin family. It acts as a neurotransmitter and neuromodulator. It is closely related to neurokinin A, both originating from the same precursor preprotachykinin A. It is released from the terminals of sensory nerves and is involved in inflammation and pain processes. This peptide is a fluorescent (FAM)-labeled Substance P, Abs/Em = 494/521 nm.
Sequence:FAM-RPKPQQFFGLM-NH2
MW:1706 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...