Cyclo (-RGDyK)
Supplier: Anaspec Inc
AS-61183-5
103006-396EA
363.92
USD
103006-396
Cyclo (-RGDyK)
Proteins and Peptides
This cyclic RGD peptide contains a subsituted d-Tyr instead of d-Phe on the fourth position. Substitution with Tyr results in a high affinity and selectivity for the avb3 integrin. Substitution with Tyr also allows for electrophilic radiohalogenation if desired.
Sequence:Cyclo(-RGDyK)
MW:619.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...