Human Ang II TAMRA peptide, TAMRA (5-Carboxytetramethylrhodamin)
Supplier: Anaspec Inc
AS-61181
103006-390EA
537.85
USD
103006-390
Human Ang II TAMRA peptide, TAMRA (5-Carboxytetramethylrhodamin)
Proteins and Peptides
This is a fluorescent (TAMRA)-labeled Angiotensin II, Abs/Em = 541/565 nm.
Sequence: TAMRA-DRVYIHPF
MW: 1458.7 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...