Simian virus V5 Epitope Tag
Supplier: Anaspec Inc
AS-61176
103006-384EA
169.54
USD
103006-384
Simian virus V5 Epitope Tag
Proteins and Peptides
This sequence is from the C-terminal sequence of the P and V proteins of Simian Virus 5.
Sequence:GKPIPNPLLGLDST
MW:1421.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...