Cyclo (-RGDfK)
Supplier: Anaspec Inc
AS-61111
103006-354EA
182.69
USD
103006-354
Cyclo (-RGDfK)
Proteins and Peptides
In one study where this peptide was labeled with 125I, it was found to bind specifically and with high affinity to alpha-v/beta-3 receptors on neovascular blood vessel sections of different major human cancers. The integrin alpha(IIb)beta(3)-specific cyclic hexapeptide contains an Arg-Gly-Asp (RGD) sequence.
Sequence:Cyclo(-RGDfK)
MW:603.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...