Cyclo (-GRGDSP) Peptide
Supplier: Anaspec Inc
AS-61110
AS-61110-5
103006-350EA
124.23
USD
103006-350
103006-352
Cyclo (-GRGDSP) Peptide
Proteins and Peptides
This cyclic peptide is a potent vasodilator. It is more powerful than the linear GRGDSP in changing the vascular tone of arterioles isolated from rat cremaster muscle.
Sequence:GRGDSP, N to C cyclized
MW:569.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...