[Ala13]-Apelin-13
Supplier: Anaspec Inc
AS-61078
103006-336EA
182.69
USD
103006-336
[Ala13]-Apelin-13
Proteins and Peptides
[Ala13]-Apelin-13 is an Apelin-13 antagonist evidenced from its blood pressure lowering reversal effects in hypertensive rats.
Sequence:QRPRLSHKGPMPA
MW:1474.7 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...