Human;Bovine bFGF (119-126)
Supplier: Anaspec Inc
AS-61070
103006-334EA
157.85
USD
103006-334
Human;Bovine bFGF (119-126)
Proteins and Peptides
This peptide corresponds to human, bovine (119-126), mouse, rat (118-125) and Heparin-Binding Growth Factor 2 (118-125) residues of bFGF. It inhibits dimerization and activation of bFGF receptors.
Sequence:KRTGQYKL
MW:993.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...