Tyrosinase-Related Protein 2 (180-188)
Supplier: Anaspec Inc
AS-61058
103006-330EA
187.08
USD
103006-330
Tyrosinase-Related Protein 2 (180-188)
Proteins and Peptides
This peptide is derived from tyrosinase-related protein 2 (TRP2) residues 180-188. TRP2 belongs to the melanocyte differentiation antigens and has been implicated as a target for immunotherapy of human as well as murine melanoma. Studies show that this TRP2 derived peptide can bind to mouse and human MHC class I molecules. Immunization with TRP2 peptide loaded dendritic cells (DCs) results in effective induction of antitumor immunity.
Sequence:SVYDFFVWL
MW:1175.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...