Mouse Uty HY Peptide (246-254)
Supplier: Anaspec Inc
AS-61045
103006-286EA
169.54
USD
103006-286
Mouse Uty HY Peptide (246-254)
Proteins and Peptides
This T-cell epitope WMHHNMDLI is a male-specific transplantation antigen, H-Y, derived from mouse UTY (ubiquitously transcribed tetratricopeptide repeat gene on the Y chromosome) protein. It is presented by H2-Db
Sequence:WMHHNMDLI
MW:1196.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...