CEF Control Peptide Pool
Supplier: Anaspec Inc
AS-61036-025
103006-276EA 1251.08 USD
103006-276
CEF Control Peptide Pool
Proteins and Peptides
This product contains 0.25 mg of the 32 CEF peptides for a total of 8 mg in one vial. Used in the stimulation of IFNγ release from CD8+ T cells in individuals with defined HLA types, these epitopes are useful in applications such as ELISPOT, intracellular cytokine and CTL assays.
% peak area by HPLC: ≥ 95
Storage condition: -20°C