Human Histatin-5
Supplier: Anaspec Inc
AS-61001
103006-240EA
363.92
USD
103006-240
Human Histatin-5
Proteins and Peptides
Histatin 5 is a human basic salivary anti-microbial peptide with strong fungicidal properties.
Sequence:DSHAKRHHGYKRKFHEKHHSHRGY
MW:3036.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...