Human BAD (103-127)
Supplier: Anaspec Inc
AS-60984
103006-120EA
380
USD
103006-120
Human BAD (103-127)
Proteins and Peptides
This is a BCL2-antagonist of cell death peptide fragment.
Sequence:NLWAAQRYGRELRRMSDEFVDSFKK
MW:3103.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...