Human Beta-Amyloid (1-42)
Supplier: Anaspec Inc
This peptide prepared by neutralizing the TFA salt form of Aß (1-42) with a dilute sodium hydroxide solution has superior solubility and fibrillogenesis properties, and the fibrils are equally neurotoxic.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 4514.1+23 Da
Molecular Weight: 4514.1 + 23
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Luo, W. et al. (2011). Synthesis and biological evaluation of novel N,N′-bis-methylenedioxybenzyl-alkylenediamines as bivalent anti-Alzheimer disease ligands. J Enzyme Inhibition Med Chem doi:10.3109/14756366.2010.548329.
Nakagami,Y. et al. (2002). A novel β-sheet breaker, RS-0406, reverses amyloid β-induced cytotoxicity and impairment of long-term potentiation in vitro. Br J Pharmacol 137, 676. doi: 10.1038/sj.bjp.0704911.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...