LSKL, Inhibitor of Thrombospondin
Supplier: Anaspec Inc
AS-60877
103006-080EA
134.46
USD
103006-080
LSKL, Inhibitor of Thrombospondin
Proteins and Peptides
This peptide, derived from the latency-associated peptide, inhibits thrombospondin (TSP-1) activation of TGF-ß; thus preventing the progression of hepatic damage and fibrosis.
Sequence:LSKL-NH2
MW:458.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...