Human Beta-Amyloid (1-42), FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-60892
103006-064EA
454.54
USD
103006-064
Human Beta-Amyloid (1-42), FAM (Carboxyfluorescein)
Proteins and Peptides
This is a fluorescent 5-FAM labeled scrambled Beta-Amyloid peptide, Abs/Em=494/521 nm.
Sequence: 5-FAM-AIAEGDSHVLKEGAYMEIFDVQGHVFGGKIFRVVDLGSHNVA
MW: 4873.4
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citation:
Shaked, BM. The FASEB Journal. 208, 1254 (2006).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...