Human sHNG
Supplier: Anaspec Inc
AS-60887
103006-054EA
415.08
USD
103006-054
Human sHNG
Proteins and Peptides
A more potent suppressor of neuronal cell death than humanin (HN), 10nM of this Gly14 substituted HN blocked cytotoxicity compared to 10uM of HN.
Sequence:MAPRGFSCLLLLTGEIDLPVKRRA
MW:2657.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...