Human ACE Substrate 2
Supplier: Anaspec Inc
AS-60757
103006-002EA
258.69
USD
103006-002
Human ACE Substrate 2
Proteins and Peptides
An ACE-2 (Angiotensin I-converting enzyme 2) fluorescent substrate. Complete hydrolysis of 0.04 mM results in a 300-fold fluorescence increase over background. Max Abs/Em=325/393 nm upon cleavage of substrate.
Sequence: Mca-APK(Dnp)
MW: 696.7 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...