Caspase 1 Inhibitor II
Supplier: Anaspec Inc
AS-60840
103005-984EA
134.46
USD
103005-984
Caspase 1 Inhibitor II
Proteins and Peptides
Caspase 1 Inhibitor II is an irreversible caspase-1, (IL-1β-converting enzyme, ICE) inhibitor. It is neuroprotective, rescuing neuronal cell cultures from cell death. It alco reduces lipopolysaccharide-lethality in animals.
Sequence:Ac-YVAD-CMK
MW:541 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...