Human Amylin
Supplier: Anaspec Inc
AS-60804
103005-980EA
580.23
USD
103005-980
Human Amylin
Proteins and Peptides
This peptide is a major constituent of protein deposits identified in the Islets of Langerhans of patients with noninsulin-dependent diabetes mellitus.
Sequence: KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY (Disulfide bridge: 2 - 7)
MW: 3904.5 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...