Human ACE Substrate 2
Supplier: Anaspec Inc
AS-60758
103005-964EA
445.77
USD
103005-964
Human ACE Substrate 2
Proteins and Peptides
Cleavage of this FRET substrate generates the fluorescent Mca signal that is monitored fluorimetrically at 393 nm, with excitation at 325 nm. A highly fluorescent substrate for caspase 1.
Sequence: Mca-YVADAPK(Dnp)
MW: 1145.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...