520 MMP FRET Substrate XIV, QXL™ 520-FAM, AnaSpec
Supplier: Anaspec Inc
Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
This peptide is a sensitive substrate for assaying MMP-1, 2, 3, 7, 8, 9, 12, and 13 activities, Abs/Em = 494/521 nm.
Sequence:QXL™ 520 -γ-Abu-P-Cha-Abu-Smc-HA-Dab(5-FAM)-AK-NH2 (Smc=S-Methyl-L-cysteine)
MW:1913.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...