Pigeon PCC (88-104)
Supplier: Anaspec Inc
AS-60432-1
103005-928EA
251.38
USD
103005-928
Pigeon PCC (88-104)
Proteins and Peptides
PCC (88-104) is a 17-mer peptide fragment of pigeon cytochrome c that stimulates proliferative T cell responses.
Sequence:KAERADLIAYLKQATAK
MW:1890.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...