Human Recombinant beta-Synuclein (from E. coli)
Supplier: Anaspec Inc
AS-55458-1000 AS-55458-500
103005-052EA 676.69 USD
103005-052 103005-054
Human Recombinant beta-Synuclein (from E. coli)
Proteins and Peptides
The sequence (Accession # NP_003076)) corresponds to the human β-synuclein along with a 6x His tag expressed in E. coli. This human β-synuclein recombinant protein is offered at a low endotoxin level of <0.1 EU per μg protein.
Beta-synuclein belongs to the family of highly conservative proteins in vertebrates. N-terminal of β-synuclein is highly homologous to α-, γ-synucleins and consists of degenerative “KTKEGV” repeats. Similar to α-synuclein, beta-synuclein is found primarily in the brain; however, it does not associate with Lewy bodies in Parkinson disease like α-synuclein. Beta-synuclein was found to inhibit production of phosphatidic acid by the phospholipase D2 transmembrane protein in vitro. In addition, beta-synuclein was detected in many breast and ovarian tumors. Recent investigations demonstrated that beta-synuclein can induce mild experimental autoimmune encephalomyelitis (EAE) in Lewis rats.
The human β-synuclein recombinant proteins from AnaSpec are offered with low endotoxin levels of <1 EU per μg protein.