EGF Receptor Substrate 2 [DADE-pY-LIPQQG], FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-29971-5
103004-432EA
2240.54
USD
103004-432
EGF Receptor Substrate 2 [DADE-pY-LIPQQG], FAM (Carboxyfluorescein)
Proteins and Peptides
The unlabeled phosphopeptide is an excellent substrate for mammalian protein tyrosine phosphatase 1B (Km = 3.9 µM) and Yersinia PTP. The sequence is derived from an autophosphorylation site (Tyr992) of EGFR. The unlabeled peptide is used to assay PTB based on marked fluorescence increase upon the removal of the phosphate group (Em = 305 nm and Ex = 282 nm).
Sequence:5-FAM-DADE-pY-LIPQQG
MW:1686.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...