Human PLP
Supplier: Anaspec Inc
AS-29213
103004-226EA
144.69
USD
103004-226
Human PLP
Proteins and Peptides
This serine substituted PLP (139-151) causes severe, acute experimental allergic encephalomyelitis in SJL mice.
Sequence: HSLGKWLGHPDKF
MW: 1521.8 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...