Human MBP (87-99)
Supplier: Anaspec Inc
AS-29211
103004-224EA
339.08
USD
103004-224
Human MBP (87-99)
Proteins and Peptides
This is fragment of the myelin basic protein (MBP), which corresponds to amino acids 84-96 of the murine sequence (86-98 in guinea pig; 87-99 in human).
Sequence:VHFFKNIVTPRTP
MW:1555.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...