c-Myc peptide epitope
Supplier: Anaspec Inc
AS-20590
103003-898EA
194.38
USD
103003-898
c-Myc peptide epitope
Proteins and Peptides
c-Myc, the product of the c-myc proto-oncogene, is a helix-loop-helix leucine zipper phosphoprotein that regulates gene transcription in cell proliferation, cell differentiation and apoptosis. This peptide is a human c-myc epitope.
Sequence:EQKLISEEDL
MW:1203.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...