Human LC-Beta-Amyloid (1-40),Biotin
Supplier: Anaspec Inc
AS-24645 AS-24645-01 AS-24648
102999-820EA 1107.85 USD
102999-820 103003-796 103003-798
Human LC-Beta-Amyloid (1-40),Biotin
Proteins and Peptides
This biotinylated Aß(1-40) contains a 6-carbon long chain (LC) to provide more accesibility for avidin attachment.
  • Peptide Content: ≥ 60%