Bovine;Human;Mouse;Rat Orexin A
Supplier: Anaspec Inc
AS-24470
103003-738EA
895.92
USD
103003-738
Bovine;Human;Mouse;Rat Orexin A
Proteins and Peptides
This hypothalamic neuropeptide has been shown to regulate feeding behavior.
Sequence:Pyr-PLPDCCRQKTCSCRLYELLHGAGNHAAGILTL-NH2 (Disulfide bridge: 6-12 and 7-14)
MW:3561.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...