Human;Pig;Rat Viasoactive intestinal peptide (1-12)

Supplier: Anaspec Inc

AS-24217
103003-696EA 118.38 USD
103003-696
Human;Pig;Rat Viasoactive intestinal peptide (1-12)
Proteins and Peptides

VIP (1−12), vasoactive intestinal peptide fragment 1-12 with a mass of 1425.5 da, is mostly used as a standard in mass spectrometry
Sequence:HSDAVFTDNYTR
MW:1425.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR