Human Gastrin Releasing Peptide
Supplier: Anaspec Inc
AS-24213
AS-24214
103003-692EA
225.08
USD
103003-692
103003-694
Human Gastrin Releasing Peptide
Proteins and Peptides
Gastrin-releasing peptide, a 27-amino acid peptide isolated from the gut, stimulates the release of Gastrin, and shares a common C-terminal decapeptide homology with bombesin. Gastrin-releasing peptide is an important growth-modulating factor in developing lung epithelium. It is used as a tumor marker in the diagnosis of small-cell lung carcinoma, since it is known to be produced by these cancer cells.
Sequence: VPLPAGGGTVLTKMYPRGNHWAVGHLM-NH2
MW: 2859.4 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...