7-Amino-4-methylcoumarin

Supplier: Anaspec Inc

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

AS-23482 AS-23483
103003-516EA 84.77 USD
103003-516 103003-518
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin

7-Amino-4-methylcoumarin is used to produce fluorogenic substrates


  • Spectral properties: Abs/Em = 351/430 nm
  • Appearance: Solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
Storage Temperature: Freezer
MDL Number: MFCD00006868
CAS Number: 26093-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR