Epitope tag
Supplier: Anaspec Inc
AS-60738
103003-362EA
363.92
USD
103003-362
Epitope tag
Proteins and Peptides
This epitope tag is a short hydrophilic, highly charged peptide. It is the most widely used epitope tag employed in structural and functional studies of proteins.
Sequence:DYKDDDDK
MW:1013 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...