Human STAL-2
Supplier: Anaspec Inc
AS-60679
103003-344EA
184.15
USD
103003-344
Human STAL-2
Proteins and Peptides
This hexapeptide, referred to as STAL-2 or TRAP (Thrombin Receptor Activating Peptide), is a protease-activated receptor (PAR-1) agonist peptide.
Sequence:SFLLRN-NH2
MW:747.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...