Human TET 830 Modified
Supplier: Anaspec Inc
AS-60622
103003-274EA
304
USD
103003-274
Human TET 830 Modified
Proteins and Peptides
TET 830 modified/T-helper epitope from tetanus toxoid is a universal human tetanus toxin T cell epitope. It induces T-cell activation and is used as a helper peptide in vaccinations.
Sequence:AQYIKANSKFIGITEL
MW:1797.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...