MUC1
Supplier: Anaspec Inc
AS-60600-1
103003-254EA
258.69
USD
103003-254
MUC1
Proteins and Peptides
This is a the tandem repeat sequence of the MUC1 mucin core. MUC1 is a high molecular weight glycoprotein over-expressed by adenocarcinomas, and by different types of bladder tumors. It is also expressed by normal human epithelial cells.
Sequence:APDTRPAPG
MW:1484.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...