NDP-MSH, 5-TAMRA-labeled, TAMRA (5-Carboxytetramethylrhodamin)
Supplier: Anaspec Inc
AS-60564-1
103003-236EA
685.46
USD
103003-236
NDP-MSH, 5-TAMRA-labeled, TAMRA (5-Carboxytetramethylrhodamin)
Proteins and Peptides
This NDP-MSH peptide is fluorescently (5-TAMRA)-labeled on the N-terminus. Abs/Em=544/572 nm. NDP-MSH is a synthetic analog of alpha-Melanotropin (alpha-MSH, alpha-melanocyte-stimulating hormone) in which Asp4 has been replaced with Norleucine and L-Phe with the D-isomer of Phe [Nle4D-Phe7] (NDP). NDP-MSH is a more potent in increasing pigmentation and decreasing UV damage in fair-skinned Caucasian volunteers.
Sequence: 5-TMR-SYS-Nle-EHfRWGKPV-NH2
MW: 2017.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...