Human Beta-Amyloid (1-42), HiLyte Fluor® 555
Supplier: Anaspec Inc
This is a fluorescent (HiLyte™ Fluor 555)-labeled ß-Amyloid peptide, Abs/Em=551/567 nm.
Sequence: HiLyte™ Fluor 555-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 5366.57 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Condello, C. et al. (2015). Microglia constitute a barrier that prevents neurotoxic protofibrillar Aβ42 hotspots around plaques. Nat Commun doi:10.1038/ncomms7176.
Epelbaum, S. et al. (2015). Acute amnestic encephalopathy in amyloid-ß oligomers injected mice is due to their widespread diffusion in vivo. Neurobiol Aging doi: 10.1016/j.neurobiolaging.2015.03.005.
Sonkar, VK. et al. (2014). Amyloid β peptide stimulates platelet activation through RhoA-dependent modulation of actomyosin organization. FASEB J 28, 1819. doi: 10.1096/fj.13-243691.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...