Human Beta-Amyloid (1-42), HiLyte Fluor® 488
Supplier: Anaspec Inc
This is a fluorescent (HiLyte™ Fluor 488)-labeled ß-Amyloid peptide, Abs/Em=503/528 nm. Hilyte 488™ Fluor labeled Aß (1-42) has a brighter intensity than FAM-labeled Aß (1-42).
Sequence: HiLyte™ Fluor 488-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 4870.5 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Minogue, A. et al. (2015). Bone marrow-derived macrophages from AβPP/PS1 mice are sensitized to the effects of inflammatory stimuli. J Alz Dis 44, 949. doi: 10.3233/JAD-142076.
Esbjörner, EK. et al. (2014). Direct observations of Amyloid β self-assembly in live cells provide insights into differences in the kinetics of Aβ (1–40) and Aβ (1–42) aggregation. Chemistry Biol 21, 732. doi:10.1016/j.chembiol.2014.03.014.
Mittag, JJ. et al. (2014). Simultaneous measurement of a range of particle sizes during Aβ < sub> 1–42 fibrillogenesis quantified using fluorescence correlation spectroscopy. Biochem Biophys Res Commun 448, 195. doi: 10.1016/j.bbrc.2014.04.088.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...