Human Amylin
Supplier: Anaspec Inc
AS-60259-1
103003-054EA
146.15
USD
103003-054
Human Amylin
Proteins and Peptides
This core region of human amylin, corresponding to amino acid residues 20 to 29, forms amyloid-like fibrils that display polymorphic structures.
Sequence: SNNFGAILSS
MW: 1009.1 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...