Chicken OVA (257-264)
Supplier: Anaspec Inc
AS-60193-1
AS-60193-5
103002-842EA
115.46
USD
103002-842
103002-988
Chicken OVA (257-264)
Proteins and Peptides
This is a class I (Kb)-restricted peptide epitope of OVA, an octameric peptide from ovalbumin presented by the class I MHC molecule, H-2Kb.
Sequence: SIINFEKL
MW: 963.2 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...