Mouse;Rat Myelin oligodendrocyte glycoprotein

Supplier: Anaspec Inc

AS-60130-1 AS-60130-10 AS-60130-5
103002-980EA 279.15 USD
103002-980 103002-982 103002-984
Mouse;Rat Myelin oligodendrocyte glycoprotein
Proteins and Peptides

Myelin oligodendrocyte glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system. It is a glycoprotein observed to be important in the myelination of nerves. It is a central molecule actively studied for its role in Multiple Sclerosis. MOG (35-55) is able to induce autoantibody production and relapsing-remitting neurological disease causing extensive plaque-like demyelination. Autoantibody response to MOG (35-55) has been observed in multiple sclerosis (MS) patients and MOG (35-55)-induced experimental autoimmune encephalomyelitis (EAE) C57/BL6 mice and Lewis rats.
Sequence: MEVGWYRSPFSRVVHLYRNGK
MW: 2582 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR