NGR Peptide 1
Supplier: Anaspec Inc
AS-60189-1
103002-836EA
454.54
USD
103002-836
NGR Peptide 1
Proteins and Peptides
This pro-apoptotic peptide, also called (klaklak)2, is a programmed cell-death inducing peptide, non toxic outside cells, but toxic once internalized, as it disrupts mitochondiral membranes. It is linked to a targeting domain (NGR), which drives the peptide to target cells and allows its internalization. It contains the Asn-Gly-Arg (NGR) motif which was shown to allow the delivery of anti-tumor compounds to tumor vessels.
Sequence:CNGRCGGklaklakklaklak-NH2 (Disulfide bridge: 1-5)
MW:2168.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...